Assay Details
| Gene Symbol: sult2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126553.1 | NP_001120025.1 | 7 - 8 | 736 | 120 | |
| XM_018095256.1 | XP_017950745.1 | 6 - 7 | 715 | 120 | ||
| GenBank mRNA | BC158326.1 | - | 7 - 8 | 736 | 120 | |
| CT025498.2 | - | 6 - 7 | 666 | 120 | ||
More Information
Set Membership: |
|

sult2a1
Genomic Map