Assay Details
| Gene Symbol: rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126557.1 | NP_001120029.1 | 2 - 3 | 628 | 88 | |
| XM_012955646.1 | XP_012811100.1 | 3 - 4 | 483 | 88 | ||
| GenBank mRNA | BC135694.1 | - | 2 - 3 | 161 | 88 | |
| BC158335.1 | - | 2 - 3 | 628 | 88 | ||
| BC168113.1 | - | 2 - 3 | 156 | 88 | ||
More Information
Set Membership: |
|

rpl10a
Genomic Map