Assay Details
| Gene Symbol: scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126615.2 | NP_001120087.1 | 635 | 82 | ||
| GenBank mRNA | BC158528.1 | - | 639 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scpep1| Gene Symbol: scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126615.2 | NP_001120087.1 | 635 | 82 | ||
| GenBank mRNA | BC158528.1 | - | 639 | 82 | ||
Set Membership: |
|