Assay Details
| Gene Symbol: nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126665.1 | NP_001120137.1 | 6 - 7 | 409 | 79 | |
| XM_012964286.2 | XP_012819740.1 | 6 - 7 | 945 | 79 | ||
| XM_012964287.2 | XP_012819741.1 | 6 - 7 | 965 | 79 | ||
| GenBank mRNA | BC159082.1 | - | 6 - 7 | 409 | 79 | |
More Information
Set Membership: |
|

nt5c3a
Genomic Map