Assay Details
| Gene Symbol: wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126726.1 | NP_001120198.1 | 5 - 6 | 1038 | 69 | |
| XM_018090799.1 | XP_017946288.1 | 6 - 7 | 1481 | 69 | ||
| GenBank mRNA | BC159319.1 | - | 5 - 6 | 1038 | 69 | |
More Information
Set Membership: |
|

wsb2
Genomic Map