Assay Details
| Gene Symbol: dtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126814.1 | NP_001120286.1 | 4 - 5 | 502 | 63 | |
| GenBank mRNA | BC160555.1 | - | 4 - 5 | 502 | 63 | |
More Information
Set Membership: |
|

dtd1
Genomic Map