Assay Details
Gene Symbol: mfsd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126822.1 | NP_001120294.1 | 4 - 5 | 805 | 66 | |
XM_012966067.2 | XP_012821521.1 | 4 - 5 | 894 | 66 | ||
GenBank mRNA | BC160589.1 | - | 4 - 5 | 805 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
