Assay Details
| Gene Symbol: mfsd13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126822.1 | NP_001120294.1 | 5 - 6 | 931 | 99 | |
| XM_012966067.2 | XP_012821521.1 | 5 - 6 | 1020 | 99 | ||
| GenBank mRNA | BC160589.1 | - | 5 - 6 | 931 | 99 | |
More Information
Set Membership: |
|

mfsd13a
Genomic Map