Assay Details
| Gene Symbol: ntan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126824.1 | NP_001120296.1 | 5 - 6 | 775 | 78 | |
| XM_012970084.2 | XP_012825538.1 | 5 - 6 | 1021 | 78 | ||
| GenBank mRNA | BC160593.1 | - | 5 - 6 | 775 | 78 | |
More Information
Set Membership: |
|

ntan1
Genomic Map