Assay Details
| Gene Symbol: tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126895.1 | NP_001120367.1 | 7 - 8 | 984 | 63 | |
| XM_012956483.1 | XP_012811937.1 | 7 - 8 | 1180 | 63 | ||
| GenBank mRNA | BC135860.1 | - | 8 - 9 | 1125 | 63 | |
| BC161057.1 | - | 7 - 8 | 984 | 63 | ||
More Information
Set Membership: |
|

tbl1x
Genomic Map