Assay Details
| Gene Symbol: igf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126946.1 | NP_001120418.1 | 2 - 3 | 310 | 74 | |
| GenBank mRNA | BC161167.1 | - | 2 - 3 | 310 | 74 | |
| CR848377.2 | - | 363 | 74 | |||
More Information
Set Membership: |
|

igf3
Genomic Map