Assay Details
| Gene Symbol: gad1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126954.1 | NP_001120426.1 | 13 - 14 | 1403 | 117 | |
| XM_012964298.2 | XP_012819752.1 | 13 - 14 | 1519 | 117 | ||
| GenBank mRNA | BC161181.1 | - | 13 - 14 | 1403 | 117 | |
More Information
Set Membership: |
|

gad1.2
Genomic Map