Assay Details
| Gene Symbol: pnlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126966.1 | NP_001120438.1 | 8 - 9 | 1170 | 88 | |
| XM_018090339.1 | XP_017945828.1 | 10 - 11 | 1831 | 88 | ||
| GenBank mRNA | BC161201.1 | - | 8 - 9 | 1170 | 88 | |
| BC161786.1 | - | 9 - 10 | 1077 | 88 | ||
More Information
Set Membership: |
|

pnlip
Genomic Map