Assay Details
Gene Symbol: sirt3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127057.1 | NP_001120529.1 | 5 - 6 | 801 | 83 | |
XM_012958604.2 | XP_012814058.1 | 5 - 6 | 818 | 83 | ||
XM_018091937.1 | XP_017947426.1 | 5 - 6 | 892 | 83 | ||
GenBank mRNA | CR942482.2 | - | 5 - 6 | 801 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
