Assay Details
| Gene Symbol: ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127061.1 | NP_001120533.1 | 2 - 3 | 186 | 59 | |
| GenBank mRNA | BC161448.1 | - | 2 - 3 | 186 | 59 | |
| CT030442.1 | - | 2 - 3 | 307 | 59 | ||
More Information
Set Membership: |
|

ets2
Genomic Map