Assay Details
| Gene Symbol: mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127074.1 | NP_001120546.1 | 9 - 10 | 2378 | 58 | |
| XM_012964312.2 | XP_012819766.1 | 9 - 10 | 5186 | 58 | ||
| GenBank mRNA | BC161476.1 | - | 9 - 10 | 2378 | 58 | |
More Information
Set Membership: |
|

mastl
Genomic Map