Assay Details
| Gene Symbol: cxorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127142.1 | NP_001120614.1 | 6 - 7 | 1235 | 82 | |
| GenBank mRNA | BC161752.1 | - | 6 - 7 | 1235 | 82 | |
More Information
Set Membership: |
|

cxorf38
Genomic Map