Assay Details
| Gene Symbol: cmtm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039741.1 | NP_001034830.1 | 3 - 4 | 529 | 84 | |
| GenBank mRNA | BC090802.1 | - | 3 - 4 | 521 | 84 | |
| BC167269.1 | - | 3 - 4 | 773 | 84 | ||
| CR855776.2 | - | 3 - 4 | 529 | 84 | ||
More Information
Set Membership: |
|

cmtm6
Genomic Map