Assay Details
| Gene Symbol: prxl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097222.2 | NP_001090691.1 | 3 - 4 | 424 | 85 | |
| GenBank mRNA | BC127374.1 | - | 3 - 4 | 323 | 85 | |
| CX802408.1 | - | 3 - 4 | 424 | 85 | ||
More Information
Set Membership: |
|

prxl2a
Genomic Map