Assay Details
| Gene Symbol: naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114217.1 | NP_001107689.1 | 7 - 8 | 1027 | 66 | |
| XM_012970521.2 | XP_012825975.1 | 8 - 9 | 1212 | 66 | ||
| GenBank mRNA | BC122059.1 | - | 7 - 8 | 1027 | 66 | |
| BC154863.1 | - | 7 - 8 | 1027 | 66 | ||
| CR760851.2 | - | 7 - 8 | 1072 | 66 | ||
More Information
Set Membership: |
|

naga
Genomic Map