Assay Details
| Gene Symbol: pgam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126512.1 | NP_001119984.1 | 2 - 3 | 439 | 74 | |
| GenBank mRNA | BC158232.1 | - | 2 - 3 | 439 | 74 | |
More Information
Set Membership: |
|

pgam1
Genomic Map