Assay Details
| Gene Symbol: isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126513.1 | NP_001119985.1 | 8 - 9 | 1204 | 92 | |
| XM_012955584.2 | XP_012811038.1 | 8 - 9 | 1159 | 92 | ||
| GenBank mRNA | BC158233.1 | - | 8 - 9 | 1204 | 92 | |
| CR761533.2 | - | 7 - 8 | 1150 | 92 | ||
More Information
Set Membership: |
|

isyna1
Genomic Map