Assay Details
| Gene Symbol: hyal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126514.1 | NP_001119986.1 | 2 - 3 | 1202 | 97 | |
| GenBank mRNA | BC158239.1 | - | 2 - 3 | 1202 | 97 | |
More Information
Set Membership: |
|

hyal2
Genomic Map