Assay Details
| Gene Symbol: spock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126515.1 | NP_001119987.1 | 5 - 6 | 623 | 65 | |
| XM_012966046.2 | XP_012821500.1 | 6 - 7 | 755 | 65 | ||
| GenBank mRNA | BC158244.1 | - | 5 - 6 | 623 | 65 | |
More Information
Set Membership: |
|

spock2
Genomic Map