Assay Details
| Gene Symbol: idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126521.1 | NP_001119993.1 | 8 - 9 | 771 | 112 | |
| XM_012954116.2 | XP_012809570.1 | 8 - 9 | 1007 | 112 | ||
| XM_012954117.2 | XP_012809571.1 | 8 - 9 | 1003 | 112 | ||
| XM_012954118.2 | XP_012809572.1 | 8 - 9 | 1013 | 112 | ||
| XM_012954119.2 | XP_012809573.1 | 7 - 8 | 941 | 112 | ||
| GenBank mRNA | BC158254.1 | - | 8 - 9 | 771 | 112 | |
More Information
Set Membership: |
|

idh3b
Genomic Map