Assay Details
| Gene Symbol: tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126531.1 | NP_001120003.1 | 2 - 3 | 345 | 84 | |
| XM_012956428.2 | XP_012811882.1 | 3 - 4 | 538 | 84 | ||
| XM_018089976.1 | XP_017945465.1 | 3 - 4 | 381 | 84 | ||
| GenBank mRNA | BC158277.1 | - | 2 - 3 | 345 | 84 | |
More Information
Set Membership: |
|

tspo
Genomic Map