Assay Details
| Gene Symbol: acot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126534.1 | NP_001120006.1 | 2 - 3 | 775 | 80 | |
| GenBank mRNA | BC158300.1 | - | 2 - 3 | 775 | 80 | |
| CU025195.1 | - | 2 - 3 | 994 | 80 | ||
More Information
Set Membership: |
|

acot2
Genomic Map