Assay Details
| Gene Symbol: sgf29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126541.1 | NP_001120013.1 | 6 - 7 | 505 | 56 | |
| GenBank mRNA | BC135838.1 | - | 5 - 6 | 643 | 56 | |
| BC158311.1 | - | 6 - 7 | 505 | 56 | ||
| CR855596.2 | - | 6 - 7 | 509 | 56 | ||
| CT485748.1 | - | 5 - 6 | 454 | 56 | ||
More Information
Set Membership: |
|

sgf29
Genomic Map