Assay Details
| Gene Symbol: trir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126585.1 | NP_001120057.1 | 1 - 2 | 758 | 64 | |
| GenBank mRNA | BC158437.1 | - | 1 - 2 | 758 | 64 | |
| CR942708.2 | - | 1 - 2 | 203 | 64 | ||
More Information
Set Membership: |
|

trir
Genomic Map