Assay Details
| Gene Symbol: trir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126585.1 | NP_001120057.1 | 2 - 3 | 836 | 65 | |
| XM_012964271.2 | XP_012819725.1 | 2 - 3 | 837 | 65 | ||
| GenBank mRNA | BC135272.1 | - | 2 - 3 | 482 | 65 | |
| BC158437.1 | - | 2 - 3 | 836 | 65 | ||
| CR942708.2 | - | 2 - 3 | 281 | 65 | ||
More Information
Set Membership: |
|

trir
Genomic Map