Assay Details
Gene Symbol: c1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126591.1 | NP_001120063.1 | 1 - 2 | 80 | 73 | |
GenBank mRNA | BC158474.1 | - | 1 - 2 | 80 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
