Assay Details
Gene Symbol: c1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126591.1 | NP_001120063.1 | 2 - 3 | 180 | 105 | |
GenBank mRNA | BC158474.1 | - | 2 - 3 | 180 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
