Assay Details
| Gene Symbol: c1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126591.1 | NP_001120063.1 | 3 - 4 | 294 | 94 | |
| GenBank mRNA | BC158474.1 | - | 3 - 4 | 294 | 94 | |
More Information
Set Membership: |
|

c1orf43
Genomic Map