Assay Details
| Gene Symbol: pak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126638.1 | NP_001120110.1 | 2 - 3 | 398 | 85 | |
| GenBank mRNA | BC158972.1 | - | 2 - 3 | 398 | 85 | |
More Information
Set Membership: |
|

pak5
Genomic Map