Assay Details
| Gene Symbol: akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126686.1 | NP_001120158.1 | 4 - 5 | 397 | 76 | |
| XM_012956458.2 | XP_012811912.1 | 5 - 6 | 632 | 76 | ||
| XM_018091087.1 | XP_017946576.1 | 4 - 5 | 591 | 76 | ||
| GenBank mRNA | BC159157.1 | - | 4 - 5 | 397 | 76 | |
| CR760988.2 | - | 5 - 6 | 569 | 76 | ||
More Information
Set Membership: |
|

akap11
Genomic Map