Assay Details
| Gene Symbol: parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126691.1 | NP_001120163.1 | 2187 | 91 | ||
| GenBank mRNA | BC159165.1 | - | 2187 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

parp9| Gene Symbol: parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126691.1 | NP_001120163.1 | 2187 | 91 | ||
| GenBank mRNA | BC159165.1 | - | 2187 | 91 | ||
Set Membership: |
|