Assay Details
| Gene Symbol: jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126752.1 | NP_001120224.1 | 1 - 2 | 633 | 69 | |
| GenBank mRNA | BC160387.1 | - | 1 - 2 | 633 | 69 | |
| CR855570.2 | - | 1 - 2 | 588 | 69 | ||
More Information
Set Membership: |
|

jph1
Genomic Map