Assay Details
| Gene Symbol: notum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126756.1 | NP_001120228.2 | 3 - 4 | 442 | 57 | |
| GenBank mRNA | BC160399.1 | - | 3 - 4 | 442 | 57 | |
More Information
Set Membership: |
|

notum
Genomic Map