Assay Details
| Gene Symbol: gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126772.2 | NP_001120244.2 | 4 - 5 | 555 | 87 | |
| XM_012966062.1 | XP_012821516.1 | 4 - 5 | 507 | 87 | ||
| XM_012966063.2 | XP_012821517.1 | 4 - 5 | 466 | 87 | ||
| XM_012966064.2 | XP_012821518.1 | 4 - 5 | 460 | 87 | ||
| XM_012966065.2 | XP_012821519.1 | 4 - 5 | 476 | 87 | ||
| GenBank mRNA | BC160440.1 | - | 3 - 4 | 436 | 87 | |
| CT030609.1 | - | 4 - 5 | 555 | 87 | ||
More Information
Set Membership: |
|

gbp5
Genomic Map