Assay Details
| Gene Symbol: upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126792.1 | NP_001120264.1 | 9 - 10 | 1070 | 70 | |
| GenBank mRNA | BC098092.1 | - | 7 - 8 | 3006 | 70 | |
| BC160511.1 | - | 9 - 10 | 1070 | 70 | ||
More Information
Set Membership: |
|

upb1
Genomic Map