Assay Details
| Gene Symbol: dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126796.1 | NP_001120268.1 | 2 - 3 | 619 | 78 | |
| GenBank mRNA | BC160517.1 | - | 2 - 3 | 619 | 78 | |
| CR942332.2 | - | 2 - 3 | 511 | 78 | ||
More Information
Set Membership: |
|

dse
Genomic Map