Assay Details
| Gene Symbol: sugt1p4-stra6lp-ccdc180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126799.1 | NP_001120271.1 | 6 - 7 | 588 | 70 | |
| XM_012955944.2 | XP_012811398.1 | 6 - 7 | 752 | 70 | ||
| GenBank mRNA | BC160523.1 | - | 6 - 7 | 588 | 70 | |
More Information
Set Membership: |
|

sugt1p4-stra6lp-ccdc180
Genomic Map