Assay Details
Gene Symbol: c6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126806.1 | NP_001120278.1 | 3 - 4 | 1027 | 90 | |
GenBank mRNA | BC160541.1 | - | 3 - 4 | 1027 | 90 | |
CU025200.1 | - | 3 - 4 | 453 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
