Assay Details
Gene Symbol: c6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126806.1 | NP_001120278.1 | 4 - 5 | 1154 | 84 | |
GenBank mRNA | BC160541.1 | - | 4 - 5 | 1154 | 84 | |
CU025200.1 | - | 4 - 5 | 580 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
