Assay Details
Gene Symbol: lrrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126807.1 | NP_001120279.1 | 2 - 3 | 812 | 94 | |
GenBank mRNA | BC160543.1 | - | 2 - 3 | 812 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
