Assay Details
| Gene Symbol: nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126918.1 | NP_001120390.1 | 699 | 62 | ||
| GenBank mRNA | BC161095.1 | - | 699 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nfkbie| Gene Symbol: nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126918.1 | NP_001120390.1 | 699 | 62 | ||
| GenBank mRNA | BC161095.1 | - | 699 | 62 | ||
Set Membership: |
|