Assay Details
| Gene Symbol: gna13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126939.1 | NP_001120411.1 | 2 - 3 | 799 | 67 | |
| GenBank mRNA | BC161136.1 | - | 2 - 3 | 799 | 67 | |
More Information
Set Membership: |
|

gna13
Genomic Map