Assay Details
| Gene Symbol: uts2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126995.1 | NP_001120467.1 | 4 - 5 | 359 | 98 | |
| GenBank mRNA | BC161294.1 | - | 4 - 5 | 359 | 98 | |
More Information
Set Membership: |
|

uts2b
Genomic Map