Assay Details
| Gene Symbol: parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127042.1 | NP_001120514.1 | 5 - 6 | 1200 | 66 | |
| XM_018095271.1 | XP_017950760.1 | 6 - 7 | 1296 | 66 | ||
| XM_018095272.1 | XP_017950761.1 | 6 - 7 | 1301 | 66 | ||
| GenBank mRNA | BC161414.1 | - | 5 - 6 | 1200 | 66 | |
More Information
Set Membership: |
|

parg
Genomic Map