Assay Details
| Gene Symbol: gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127048.1 | NP_001120520.1 | 278 | 54 | ||
| GenBank mRNA | BC161426.1 | - | 278 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gas7| Gene Symbol: gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127048.1 | NP_001120520.1 | 278 | 54 | ||
| GenBank mRNA | BC161426.1 | - | 278 | 54 | ||
Set Membership: |
|